missing translation for 'onlineSavingsMsg'
Learn More

4-Bromostyrene, 96%, stabilized, Thermo Scientific Chemicals

Catalog Number p-4510264
Click to view available options
Quantity:
10 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
2039-82-9
C8H7Br
183.05
MFCD00000110
WGGLDBIZIQMEGH-UHFFFAOYSA-N
4-bromostyrene, 1-bromo-4-vinylbenzene, p-bromostyrene, benzene, 1-bromo-4-ethenyl, styrene, p-bromo, 1-4-bromophenyl ethylene, unii-tvk4s80xlu, p-bromo styrene, 1-bromo-4-vinyl-benzene, tvk4s80xlu
16263
1-bromo-4-ethenylbenzene
BrC1=CC=C(C=C)C=C1
This item is not returnable. View return policy
2039-82-9
C8H7Br
183.05
MFCD00000110
WGGLDBIZIQMEGH-UHFFFAOYSA-N
4-bromostyrene, 1-bromo-4-vinylbenzene, p-bromostyrene, benzene, 1-bromo-4-ethenyl, styrene, p-bromo, 1-4-bromophenyl ethylene, unii-tvk4s80xlu, p-bromo styrene, 1-bromo-4-vinyl-benzene, tvk4s80xlu
16263
1-bromo-4-ethenylbenzene
BrC1=CC=C(C=C)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 4.5°C
Color Colorless to Yellow
Density 1.4000g/mL
Boiling Point 88.0°C to 89.0°C (16.0 mmHg)
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHC6H4Br
Refractive Index 1.5930 to 1.5950
Quantity 10 mL
Beilstein 05,II,367
Specific Gravity 1.4
Stabilizer 100 to 500ppm 4-tert.-butylcatechol
Formula Weight 183.05
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 4-Bromostyrene
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromostyrene, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'