missing translation for 'onlineSavingsMsg'
Learn More

4-Bromostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog Number p-7023891
Click to view available options
Quantity:
5 g
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2039-82-9
C8H7Br
183.05
MFCD00000110
WGGLDBIZIQMEGH-UHFFFAOYSA-N
4-bromostyrene, 1-bromo-4-vinylbenzene, p-bromostyrene, benzene, 1-bromo-4-ethenyl, styrene, p-bromo, 1-4-bromophenyl ethylene, unii-tvk4s80xlu, p-bromo styrene, 1-bromo-4-vinyl-benzene, tvk4s80xlu
16263
1-bromo-4-ethenylbenzene
BrC1=CC=C(C=C)C=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2039-82-9
C8H7Br
183.05
MFCD00000110
WGGLDBIZIQMEGH-UHFFFAOYSA-N
4-bromostyrene, 1-bromo-4-vinylbenzene, p-bromostyrene, benzene, 1-bromo-4-ethenyl, styrene, p-bromo, 1-4-bromophenyl ethylene, unii-tvk4s80xlu, p-bromo styrene, 1-bromo-4-vinyl-benzene, tvk4s80xlu
16263
1-bromo-4-ethenylbenzene
BrC1=CC=C(C=C)C=C1

4-Bromostyrene is widely utilized in the synthesis of nitroolefins by alkene cross-metathesis. It plays an important role in the Heck reaction to the synthesis of poly(1,4-phenylenevinylene). It is employed in the structure activity relationships (SAR) study of the chemical and biochemical properties of the vinyl group of styrene. It is also used to investigate the photochemical growth of Br-terminated self-assembled monolayers. It is also involved in the synthesis of silsesquioxanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromostyrene is widely utilized in the synthesis of nitroolefins by alkene cross-metathesis. It plays an important role in the Heck reaction to the synthesis of poly(1,4-phenylenevinylene). It is employed in the structure activity relationships (SAR) study of the chemical and biochemical properties of the vinyl group of styrene. It is also used to investigate the photochemical growth of Br-terminated self-assembled monolayers. It is also involved in the synthesis of silsesquioxanes.

Solubility
Miscible with ethanol, ether and benzene.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, bases, oxidizing agents and halogens. Keep away from sources of ignition.
TRUSTED_SUSTAINABILITY
Melting Point 3°C to 6°C
Density 1.4
Boiling Point 88°C to 89°C (16 mmHg)
Flash Point 75°C (167°F)
Refractive Index 1.594
Quantity 5 g
Beilstein 1634204
Solubility Information Miscible with ethanol,ether and benzene.
Formula Weight 183.05
Percent Purity 98%
Chemical Name or Material 4-Bromostyrene, stabilized with 0.1% 4-tert-butylcatechol
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'