Learn More
4-Bromopyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
4-Bromopicolinic acid is a useful synthetic intermediate. Also used as a intermediate for agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromopicolinic acid is a useful synthetic intermediate. Also used as a intermediate for agrochemical, pharmaceutical and dyestuff field.
Solubility
Sparingly soluble in dimethyl sulfoxide , methanol and water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
4-Bromopicolinic acid is a useful synthetic intermediate. Also used as a intermediate for agrochemical, pharmaceutical and dyestuff field.
Solubility
Sparingly soluble in dimethyl sulfoxide , methanol and water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 177°C to 179°C |
Quantity | 1 g |
Beilstein | 471891 |
Solubility Information | Sparingly soluble in dimethyl sulfoxide ,methanol and water. |
Formula Weight | 202.01 |
Percent Purity | 97% |
Chemical Name or Material | 4-Bromopyridine-2-carboxylic acid |