missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)-3-methyl-5-phenylisoxazole, ≥95%, Thermo Scientific™

Catalog Number p-2700742
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
113841-59-1
C11H10BrNO
252.111
MFCD03086124
KCGVZUOBUPWFJC-UHFFFAOYSA-N
2776518
4-(bromomethyl)-3-methyl-5-phenyl-1,2-oxazole
CC1=NOC(=C1CBr)C2=CC=CC=C2
This item is not returnable. View return policy
113841-59-1
C11H10BrNO
252.111
MFCD03086124
KCGVZUOBUPWFJC-UHFFFAOYSA-N
2776518
4-(bromomethyl)-3-methyl-5-phenyl-1,2-oxazole
CC1=NOC(=C1CBr)C2=CC=CC=C2
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 4-bromomethyl-3-methyl-5-phenylisoxazole, 4-bromomethyl-3-methyl-5-phenyl-1,2-oxazole, pubchem23336, acmc-1c8sr, 4-bromomethyl-3-methyl-5-phenyl-isoxazole, isoxazole,4-bromomethyl-3-methyl-5-phenyl, isoxazole, 4-bromomethyl-3-methyl-5-phenyl
Formula Weight 252.11
Percent Purity 95%
Chemical Name or Material 4-(Bromomethyl)-3-methyl-5-phenylisoxazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'