Learn More
Alfa Aesar™ 4-Bromo-N,N-dicyclohexylbenzamide, 97%
Brand:  Alfa Aesar™ H58734.MC
Additional Details : CAS Number : 327058-58-2 Weight : 0.00010kg
Quantity 100mg
 View more versions of this product
Product Code. 15484948
  • 108.00 EUR / 100mg
Estimated Shipment date
from Supplier 06-08-2018
Add to basket
Specifications
Chemical Identifiers
327058-58-2
364.327
CFPQPOGTCPLGSE-UHFFFAOYSA-N
C19H26BrNO
MFCD00585206
3604492
Specifications
4-Bromo-N,N-dicyclohexylbenzamide
327058-58-2
100mg
C19H26BrNO
CFPQPOGTCPLGSE-UHFFFAOYSA-N
4-bromo-N,N-dicyclohexylbenzamide
3604492
364.32
Solid
97%
MFCD00585206
C1CCC(CC1)N(C2CCCCC2)C(=O)C3=CC=C(C=C3)Br
364.327