missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-cyclohexyl-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62143.03

Additional Details : CAS Number : 1020253-02-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15487076

  • 323.00 EUR / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-cyclohexyl-3-(trifluoromethyl)benzenesulfonamide
1020253-02-4
386.23
MFCD09972158

Specifications

4-Bromo-N-cyclohexyl-3-(trifluoromethyl)benzenesulfonamide
Solid
97%
MFCD09972158
1020253-02-4
1g
386.23
Videos
SDS