missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(4-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63654.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15458426

  • 1291.00 EUR / 5g


Estimated Shipment date
from Supplier 19-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD26143379

Specifications

4-Bromo-N-(4-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide
MFCD26143379
Solid
5g
410.21
97%
Videos
SDS