Learn More
Alfa Aesar™ 4-Bromo-N-[4-fluoro-3-(trifluoromethyl)phenyl]-3-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H63230.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15485595
  • 1291.00 EUR / 5g
Estimated Shipment date
from Supplier 05-02-2018
Add to basket
Specifications
Chemical Identifiers
4-Bromo-N-[4-fluoro-3-(trifluoromethyl)phenyl]-3-(trifluoromethyl)benzenesulfonamide
466.17
MFCD26143355
Specifications
4-Bromo-N-[4-fluoro-3-(trifluoromethyl)phenyl]-3-(trifluoromethyl)benzenesulfonamide
97%
466.17
Solid
5g
MFCD26143355