missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(4-chlorobenzyl)-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64968.03

Additional Details : CAS Number : 1350926-49-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459286

  • 285.00 EUR / 1g


Estimated Shipment date
from Supplier 24-06-2019
Add to basket

Specifications

Chemical Identifiers

1350926-49-6
378.64
ITAJJFKCDMJPEJ-UHFFFAOYSA-N
98000860
C1=CC(=CC=C1CNS(=O)(=O)C2=C(C=C(C=C2)Br)F)Cl
C13H10BrClFNO2S
MFCD28098734
n-4-chlorobenzyl-4-bromo-2-fluoro-benzenesulfonamide, 4-bromo-n-4-chlorophenyl methyl-2-fluorobenzenesulfonamide
4-bromo-N-[(4-chlorophenyl)methyl]-2-fluorobenzenesulfonamide

Specifications

4-Bromo-N-(4-chlorobenzyl)-2-fluorobenzenesulfonamide
1350926-49-6
MFCD28098734
ITAJJFKCDMJPEJ-UHFFFAOYSA-N
4-bromo-N-[(4-chlorophenyl)methyl]-2-fluorobenzenesulfonamide
98000860
Solid
1g
C13H10BrClFNO2S
n-4-chlorobenzyl-4-bromo-2-fluoro-benzenesulfonamide, 4-bromo-n-4-chlorophenyl methyl-2-fluorobenzenesulfonamide
C1=CC(=CC=C1CNS(=O)(=O)C2=C(C=C(C=C2)Br)F)Cl
378.64
378.64
97%
Videos
SDS Documents