missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(3-bromophenyl)-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64237.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458796

  • 285.00 EUR / 1g


Estimated Shipment date
from Supplier 18-06-2019
Add to basket

Specifications

Chemical Identifiers

1772777-01-1
409.07
SWWQVDNSEJFEPC-UHFFFAOYSA-N
97455656
C12H8Br2FNO2S
MFCD27824753
4-bromo-n-3-bromophenyl-2-fluorobenzenesulfonamide

Specifications

4-Bromo-N-(3-bromophenyl)-2-fluorobenzenesulfonamide
1772777-01-1
MFCD27824753
SWWQVDNSEJFEPC-UHFFFAOYSA-N
97455656
Solid
1g
C12H8Br2FNO2S
4-bromo-n-3-bromophenyl-2-fluorobenzenesulfonamide
409.07
409.07
97%
Videos
SDS