missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-fluoro-N-(3-methylphenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64204.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458766

  • 256.00 EUR / 1g


Estimated Shipment date
from Supplier 14-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27392147
4-Bromo-2-fluoro-N-(m-tolyl)benzenesulfonamide

Specifications

4-Bromo-2-fluoro-N-(3-methylphenyl)benzenesulfonamide
MFCD27392147
344.2
97%
1g
4-Bromo-2-fluoro-N-(m-tolyl)benzenesulfonamide
Solid
Videos
SDS