missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-chloropyrimidine, 98%, Thermo Scientific™

Catalog Number 11410010
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
5305-59-9
C4H4ClN3
129.547
MFCD00053576
DUKKRSPKJMHASP-UHFFFAOYSA-N
4-amino-6-chloropyrimidine, 6-chloropyrimidin-4-ylamine, 6-chloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 6-chloro, 6-chloro-4-pyrimidinamine, 6-amino-4-chloropyrimidine, 6-chloropyrimidine-4-ylamine, 6-chloro-4-aminopyrimidine, 6-chlorpyrimidin-4-amin, zlchem 630
238012
6-chloropyrimidin-4-amine
C1=C(N=CN=C1Cl)N
This item is not returnable. View return policy
5305-59-9
C4H4ClN3
129.547
MFCD00053576
DUKKRSPKJMHASP-UHFFFAOYSA-N
4-amino-6-chloropyrimidine, 6-chloropyrimidin-4-ylamine, 6-chloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 6-chloro, 6-chloro-4-pyrimidinamine, 6-amino-4-chloropyrimidine, 6-chloropyrimidine-4-ylamine, 6-chloro-4-aminopyrimidine, 6-chlorpyrimidin-4-amin, zlchem 630
238012
6-chloropyrimidin-4-amine
C1=C(N=CN=C1Cl)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 129.55
Percent Purity 98%
Chemical Name or Material 4-Amino-6-chloropyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'