missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1H-pyrazole, 95%, Thermo Scientific Chemicals

Catalog Number p-7030465
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
28466-26-4
C3H5N3
83.094
MFCD01693729
AXINVSXSGNSVLV-UHFFFAOYSA-N
4-aminopyrazole, 4-amino-1h-pyrazole, 1h-pyrazol-4-ylamine, 9ci, pyrazole, 4-amino, 1h-pyrazole-4-amine, pyrazole-4-ylamine, 4-nh2-pyrazole, 1-h-pyrazol-4-amine, pubchem14988
78035
1H-pyrazol-4-amine
C1=C(C=NN1)N
This item is not returnable. View return policy
28466-26-4
C3H5N3
83.094
MFCD01693729
AXINVSXSGNSVLV-UHFFFAOYSA-N
4-aminopyrazole, 4-amino-1h-pyrazole, 1h-pyrazol-4-ylamine, 9ci, pyrazole, 4-amino, 1h-pyrazole-4-amine, pyrazole-4-ylamine, 4-nh2-pyrazole, 1-h-pyrazol-4-amine, pubchem14988
78035
1H-pyrazol-4-amine
C1=C(C=NN1)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 72°C to 77°C
Quantity 100 mg
Sensitivity Hygroscopic
Formula Weight 83.09
Percent Purity 95%
Chemical Name or Material 4-Amino-1H-pyrazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'