Learn More
4-Amino-1,2,4-triazole, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4-Amino-1,2,4-triazole is used as an intermediate for the preparation of antifungal agent like fluconazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Amino-1,2,4-triazole is used as an intermediate for the preparation of antifungal agent like fluconazole.
Solubility
Soluble in water and methanol.
Notes
Incompatible with strong oxidizing agents.
4-Amino-1,2,4-triazole is used as an intermediate for the preparation of antifungal agent like fluconazole.
Solubility
Soluble in water and methanol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 83°C to 88°C |
Quantity | 5 g |
Beilstein | 107563 |
Solubility Information | Soluble in water and methanol. |
Formula Weight | 84.08 |
Percent Purity | 99% |
Chemical Name or Material | 4-Amino-1,2,4-triazole |