missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4,7-Dimethyl-5-decyn-4,7-diol, (+/-) + meso, 98+%

Catalog Number 11408947
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
126-87-4
C12H22O2
198.306
MFCD00021836
HCKFFIBKYQSDRD-UHFFFAOYSA-N
135969
4,7-dimethyldec-5-yne-4,7-diol
CCCC(C)(C#CC(C)(CCC)O)O
This item is not returnable. View return policy
126-87-4
C12H22O2
198.306
MFCD00021836
HCKFFIBKYQSDRD-UHFFFAOYSA-N
135969
4,7-dimethyldec-5-yne-4,7-diol
CCCC(C)(C#CC(C)(CCC)O)O
Melting Point 67°C to 70°C
Boiling Point 125°C to 127°C (3 mmHg)
Quantity 5 g
Beilstein 1723151
Synonym 4,7-dimethyl-5-decyne-4,7-diol, 4,7-dimethyl-5-decyn-4,7-diol, surfynol 102, 5-decyne-4,7-diol,4,7-dimethyl, acmc-1byq5, 5-decyne-4, 4,7-dimethyl, 5-decyne-4,7-diol, 4,7-dimethyl, 4,7-dimethyl-5-decyn-4,7-diol, + meso
Formula Weight 198.31
Percent Purity ≥98%
Chemical Name or Material 4,7-Dimethyl-5-decyn-4,7-diol, (+/-) + meso
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'