missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-4228906
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
40133-07-1
ROLXOQXKNDKXTA-UHFFFAOYSA-N
4,5,6,7-tetrahydrobenzo b thiophene-2-carboxylic acid, 4,5,6,7-tetrahydro-benzo b thiophene-2-carboxylic acid, benzo b thiophene-2-carboxylic acid, 4,5,6,7-tetrahydro, acmc-1asbb, enamine_001826, 2-carboxy-4,5,6,7-tetrahydro-1-benzothiophene, 2-carboxy-4,5,6,7-tetrahydrobenzo b thiophene, 4,5,6,7-tetrahydrobenzo b thiophene-2-carboxylicacid, 4,5,6,7-tetrahydrobenzo b-thiophene-2-carboxylic acid, 4,5,6,7-tetrahydro-benzo b thiophene-2-carboxylic
2063443
4,5,6,7-tetrahydro-1-benzothiophene-2-carboxylic acid
C1CCC2=C(C1)C=C(S2)C(=O)O
This item is not returnable. View return policy
40133-07-1
ROLXOQXKNDKXTA-UHFFFAOYSA-N
4,5,6,7-tetrahydrobenzo b thiophene-2-carboxylic acid, 4,5,6,7-tetrahydro-benzo b thiophene-2-carboxylic acid, benzo b thiophene-2-carboxylic acid, 4,5,6,7-tetrahydro, acmc-1asbb, enamine_001826, 2-carboxy-4,5,6,7-tetrahydro-1-benzothiophene, 2-carboxy-4,5,6,7-tetrahydrobenzo b thiophene, 4,5,6,7-tetrahydrobenzo b thiophene-2-carboxylicacid, 4,5,6,7-tetrahydrobenzo b-thiophene-2-carboxylic acid, 4,5,6,7-tetrahydro-benzo b thiophene-2-carboxylic
2063443
4,5,6,7-tetrahydro-1-benzothiophene-2-carboxylic acid
C1CCC2=C(C1)C=C(S2)C(=O)O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 195°C to 205°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C9H10O2S
Quantity 1 g
Molecular Weight (g/mol) 182.237
Formula Weight 182.24
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4,5,6,7-Tetrahydrobenzo¢b!thiophene-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'