missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4,5,6,7-Tetrahydro-5-azabenzimidazole hydrochloride, 95%

Brand:  Alfa Aesar™ H64755.03

Additional Details : CAS Number : 879668-17-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459126

  • 352.00 EUR / 1g


Estimated Shipment date
from Supplier 19-02-2019
Add to basket

Specifications

Chemical Identifiers

879668-17-4
159.617
CHLFJSVTUQAGSC-UHFFFAOYSA-N
21330723
C1CNCC2=C1N=CN2.Cl
C6H10ClN3
MFCD11846200
4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hydrochloride, 4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hcl, 4,5,6,7-tetrahydro-1h-imidazo 4,5-c pyridine hydrochloride, 3h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloride, 4,5,6,7-tetrahydro-5-azabenzimidazole hydrochloride, 3h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloridyl, 4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hcl, 1h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloride, 4,5,6,7-tetrahydro-1h-imidazo 3,4-c pyridine hydrochloride, 4,5,6,7-tetrahydro-imidazo-4,5-c-pyridine hydrochloride
4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine;hydrochloride

Specifications

4,5,6,7-Tetrahydro-5-azabenzimidazole hydrochloride
879668-17-4
MFCD11846200
4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hydrochloride, 4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hcl, 4,5,6,7-tetrahydro-1h-imidazo 4,5-c pyridine hydrochloride, 3h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloride, 4,5,6,7-tetrahydro-5-azabenzimidazole hydrochloride, 3h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloridyl, 4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hcl, 1h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloride, 4,5,6,7-tetrahydro-1h-imidazo 3,4-c pyridine hydrochloride, 4,5,6,7-tetrahydro-imidazo-4,5-c-pyridine hydrochloride
C1CNCC2=C1N=CN2.Cl
159.617
159.62
95%
1g
C6H10ClN3
15312806
CHLFJSVTUQAGSC-UHFFFAOYSA-N
4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine;hydrochloride
21330723
Solid
Videos
SDS