Learn More
4,4'-Dibromo-2,2'-bipyridine, 98%, Thermo Scientific Chemicals
Catalog Number 15392052
Click to view available options
Quantity:
1 g
5 g
18511-71-2
C10H6Br2N2
313.98
KIIHBDSNVJRWFY-UHFFFAOYSA-N
4,4'-dibromo-2,2'-bipyridine, 4-bromo-2-4-bromopyridin-2-yl pyridine, 4,4'-dibromo-2,2'-bipyridyl, 2,2'-bipyridine, 4,4'-dibromo, bpbb01, acmc-1bo83, 4,4'-dibromo-2,2-bipyridine, 4,4'-bromo-2,2'-bipyridine, 4,4'-dibromo-2,2'-dipyridine, 4,4'-dibromine-2,2'-bipyridine
15569987
4-bromo-2-(4-bromopyridin-2-yl)pyridine
C1=CN=C(C=C1Br)C2=NC=CC(=C2)Br
18511-71-2
C10H6Br2N2
313.98
KIIHBDSNVJRWFY-UHFFFAOYSA-N
4,4'-dibromo-2,2'-bipyridine, 4-bromo-2-4-bromopyridin-2-yl pyridine, 4,4'-dibromo-2,2'-bipyridyl, 2,2'-bipyridine, 4,4'-dibromo, bpbb01, acmc-1bo83, 4,4'-dibromo-2,2-bipyridine, 4,4'-bromo-2,2'-bipyridine, 4,4'-dibromo-2,2'-dipyridine, 4,4'-dibromine-2,2'-bipyridine
15569987
4-bromo-2-(4-bromopyridin-2-yl)pyridine
C1=CN=C(C=C1Br)C2=NC=CC(=C2)Br
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 140.0°C to 143.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 313.98
Percent Purity 98%
Chemical Name or Material 4,4'-Dibromo-2,2'-bipyridine
compliance-icons
Product Identifier
  • 4,4'-Dibromo-2,2'-bipyridine, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.