Learn More
Alfa Aesar™ 4-(3-Nitrophenylsulfonyl)morpholine, 97%
Brand:  Alfa Aesar™ H58254.MC
Additional Details : CAS Number : 91619-33-9 Weight : 0.00010kg
Quantity 100mg
 View more versions of this product
Product Code. 15444578
  • 112.00 EUR / 100mg
Estimated Shipment date
from Supplier 24-04-2019
Add to basket
Specifications
Chemical Identifiers
91619-33-9
272.275
XCYXNNGNLAIXNM-UHFFFAOYSA-N
237475
C1COCCN1S(=O)(=O)C2=CC=CC(=C2)[N+](=O)[O-]
C10H12N2O5S
MFCD00622715
4-3-nitrophenylsulfonyl morpholine, 4-3-nitrophenyl sulfonyl morpholine, 4-3-nitro-benzenesulfonyl-morpholine, 4-3-nitrobenzenesulfonyl morpholine, 4-3-nitrophenyl sulfonyl morpholine, morpholine, 4-3-nitrophenyl sulfonyl, acmc-209rco, cbmicro_008083, 4-3-nitrophenyl sulfonylmorpholine, 4-3-nitrobenzene-1-sulfonyl morpholine
4-(3-nitrophenyl)sulfonylmorpholine
Specifications
4-(3-Nitrophenylsulfonyl)morpholine
100mg
C10H12N2O5S
4-3-nitrophenylsulfonyl morpholine, 4-3-nitrophenyl sulfonyl morpholine, 4-3-nitro-benzenesulfonyl-morpholine, 4-3-nitrobenzenesulfonyl morpholine, 4-3-nitrophenyl sulfonyl morpholine, morpholine, 4-3-nitrophenyl sulfonyl, acmc-209rco, cbmicro_008083, 4-3-nitrophenyl sulfonylmorpholine, 4-3-nitrobenzene-1-sulfonyl morpholine
C1COCCN1S(=O)(=O)C2=CC=CC(=C2)[N+](=O)[O-]
272.275
272.28
97%
Yellow
91619-33-9
MFCD00622715
XCYXNNGNLAIXNM-UHFFFAOYSA-N
4-(3-nitrophenyl)sulfonylmorpholine
237475
Solid