missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(2-Thiazolidinyl)benzoic acid, 97%

Brand:  Alfa Aesar™ H62360.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458888

  • 177.80 EUR / 1g


Estimated Shipment date
from Supplier 19-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD26960674

Specifications

4-(2-Thiazolidinyl)benzoic acid
MFCD26960674
Solid
1g
209.26
97%
Videos
SDS