missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Furyl)-2-methylthiazole, 97%, Thermo Scientific™

Catalog Number 15484458
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
492440-71-8
C8H7NOS
165.21
MFCD22682939
AGIHEIJGDJIILG-UHFFFAOYSA-N
4-2-furyl-2-methylthiazole, 4-furan-2-yl-2-methyl-1,3-thiazole, 4-furan-2-yl-2-methylthiazole
15704191
4-(furan-2-yl)-2-methyl-1,3-thiazole
CC1=NC(=CS1)C2=CC=CO2
This item is not returnable. View return policy
492440-71-8
C8H7NOS
165.21
MFCD22682939
AGIHEIJGDJIILG-UHFFFAOYSA-N
4-2-furyl-2-methylthiazole, 4-furan-2-yl-2-methyl-1,3-thiazole, 4-furan-2-yl-2-methylthiazole
15704191
4-(furan-2-yl)-2-methyl-1,3-thiazole
CC1=NC(=CS1)C2=CC=CO2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 128°C
Quantity 1 g
Beilstein 128400
Formula Weight 165.2
Percent Purity 97%
Chemical Name or Material 4-(2-Furyl)-2-methylthiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'