Learn More
4-(1H-Pyrazol-1-yl)aniline, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 159.19 |
Percent Purity | 97% |
Chemical Name or Material | 4-(1H-Pyrazol-1-yl)aniline |