Learn More
3M™ Versaflo™ Breathing Tubes for Headtops and Air Delivery Units
Lightweight BT-series breathing tubes are compatible with all 3M™ Versaflo™ headtops and air delivery units.
Brand: 3M™ 7000000158
48.95 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.23670kg
Description
- Long, lightweight breathing tube that is easy to wipe down
- Quick release swivel enables one-handed connection and release
- Disposable breathing tube cover is available to help cleaning and decontamination operations
- 3M Catalog# BT-20S and BT-20L
Specifications
Breathing Tube | |
Assembly, facility maintenance, grinding, machine operation, painting, sanding | |
BT-20S | |
71.1 cm |
Small/Medium | |
Versaflo™ PAPR Systems | |
28 in. |