missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridinesulfonic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-189465
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
636-73-7
C5H4NO3S
158.15
MFCD00006381
DVECLMOWYVDJRM-UHFFFAOYSA-M
3-pyridinesulfonic acid, pyridine-3-sulphonic acid, 3-pyridylsulfonic acid, 3-pyridine sulfonic acid, 3-sulfopyridine, beta-sulfopyridine, 3-pyridinesulfonicacid, pubchem9608, pyridin-3-sulfonic acid, acmc-209nh7
69468
pyridine-3-sulfonic acid
[O-]S(=O)(=O)C1=CC=CN=C1
This item is not returnable. View return policy
636-73-7
C5H4NO3S
158.15
MFCD00006381
DVECLMOWYVDJRM-UHFFFAOYSA-M
3-pyridinesulfonic acid, pyridine-3-sulphonic acid, 3-pyridylsulfonic acid, 3-pyridine sulfonic acid, 3-sulfopyridine, beta-sulfopyridine, 3-pyridinesulfonicacid, pubchem9608, pyridin-3-sulfonic acid, acmc-209nh7
69468
pyridine-3-sulfonic acid
[O-]S(=O)(=O)C1=CC=CN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point >300.0°C
Color White
Flash Point >66°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula (C5H4N)SO3H
Quantity 100 g
Beilstein 22, 387
Solubility Information (5% in water) Clear colorless
Formula Weight 159.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-Pyridinesulfonic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Pyridinesulfonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'