missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinecarboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2700291
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
498-95-3
C6H11NO2
129.159
MFCD00005992
XJLSEXAGTJCILF-UHFFFAOYSA-N
nipecotic acid, 3-piperidinecarboxylic acid, hexahydronicotinic acid, +/--nipecotic acid, h-dl-nip-oh, +/--3-piperidine carboxylic acid, dl-piperidine-3-carboxylic acid, nipecotic, h-nip-oh, 3-carboxypiperidine
4498
CHEBI:116931
piperidine-3-carboxylic acid
C1CC(CNC1)C(=O)O
This item is not returnable. View return policy
498-95-3
C6H11NO2
129.159
MFCD00005992
XJLSEXAGTJCILF-UHFFFAOYSA-N
nipecotic acid, 3-piperidinecarboxylic acid, hexahydronicotinic acid, +/--nipecotic acid, h-dl-nip-oh, +/--3-piperidine carboxylic acid, dl-piperidine-3-carboxylic acid, nipecotic, h-nip-oh, 3-carboxypiperidine
4498
CHEBI:116931
piperidine-3-carboxylic acid
C1CC(CNC1)C(=O)O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Beige-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 129.16
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 3-Piperidinecarboxylic acid, Functional Group amine (alkyl)
     carboxylic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'