missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropyl isothiocyanate, 97%, Thermo Scientific™

Catalog Number p-2700171
Click to view available options
Quantity:
10 g
50 g
100 g
This item is not returnable. View return policy
17702-11-3
C5H9NOS
131.193
MFCD00022063
UKVFWFDXYBLOLU-UHFFFAOYSA-N
3-methoxypropyl isothiocyanate, 3-methoxypropylisothiocyanate, 1-isothiocyanato-3-methoxy-propane, propane,1-isothiocyanato-3-methoxy, propane, 1-isothiocyanato-3-methoxy, 3-methoxypropanisothiocyanate, acmc-1bu13, 3-methoxy-n-propyl isothiocyanate, 3-methoxpropyl isothiocyanate, #
140274
1-isothiocyanato-3-methoxypropane
COCCCN=C=S
This item is not returnable. View return policy
17702-11-3
C5H9NOS
131.193
MFCD00022063
UKVFWFDXYBLOLU-UHFFFAOYSA-N
3-methoxypropyl isothiocyanate, 3-methoxypropylisothiocyanate, 1-isothiocyanato-3-methoxy-propane, propane,1-isothiocyanato-3-methoxy, propane, 1-isothiocyanato-3-methoxy, 3-methoxypropanisothiocyanate, acmc-1bu13, 3-methoxy-n-propyl isothiocyanate, 3-methoxpropyl isothiocyanate, #
140274
1-isothiocyanato-3-methoxypropane
COCCCN=C=S
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 131.2
Percent Purity 97%
Chemical Name or Material 3-Methoxypropyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'