missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(pentafluorothio)benzyl alcohol, 97%, Thermo Scientific™

Catalog Number p-7032636
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1240257-89-9
C7H6F6OS
252.174
MFCD16652458
VGHYIFHKEQFPSP-UHFFFAOYSA-N
3-fluoro-5-pentafluorosulfur benzyl alcohol, 3-fluoro-5-pentafluoro-??-sulfanyl phenyl methanol, 3-fluoro-5-pentafluorothio benzyl alcohol
66523537
[3-fluoro-5-(pentafluoro-$l^{6}-sulfanyl)phenyl]methanol
C1=C(C=C(C=C1F)S(F)(F)(F)(F)F)CO
This item is not returnable. View return policy
1240257-89-9
C7H6F6OS
252.174
MFCD16652458
VGHYIFHKEQFPSP-UHFFFAOYSA-N
3-fluoro-5-pentafluorosulfur benzyl alcohol, 3-fluoro-5-pentafluoro-??-sulfanyl phenyl methanol, 3-fluoro-5-pentafluorothio benzyl alcohol
66523537
[3-fluoro-5-(pentafluoro-$l^{6}-sulfanyl)phenyl]methanol
C1=C(C=C(C=C1F)S(F)(F)(F)(F)F)CO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 252.17
Percent Purity 97%
Chemical Name or Material 3-Fluoro-5-(pentafluorothio)benzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'