Learn More
3-Ethoxycarbonyl-4-fluorophenylboronic acid, 95%, Thermo Scientific™
Catalog Number 15259936
Click to view available options
Quantity:
250 mg
1 g
874219-36-0
C9H10BFO4
211.98
MFCD08061836
YCXMTPOFCTUKTB-UHFFFAOYSA-N
3-ethoxycarbonyl-4-fluorophenyl boronic acid, 3-ethoxycarbonyl-4-fluorobenzeneboronic acid, 3-ethoxycarbonyl-4-fluorophenylboronic acid, benzoicacid, 5-borono-2-fluoro-, 1-ethyl ester 9ci, acmc-209ql0, 3-ethoxycarbonyl-4-fluorophenylboronicacid, ksc495s8r, 4-fluoro-3-ethoxycarbonylphenylboronic acid
24901771
(3-ethoxycarbonyl-4-fluorophenyl)boronic acid
CCOC(=O)C1=C(F)C=CC(=C1)B(O)O
874219-36-0
C9H10BFO4
211.98
MFCD08061836
YCXMTPOFCTUKTB-UHFFFAOYSA-N
3-ethoxycarbonyl-4-fluorophenyl boronic acid, 3-ethoxycarbonyl-4-fluorobenzeneboronic acid, 3-ethoxycarbonyl-4-fluorophenylboronic acid, benzoicacid, 5-borono-2-fluoro-, 1-ethyl ester 9ci, acmc-209ql0, 3-ethoxycarbonyl-4-fluorophenylboronicacid, ksc495s8r, 4-fluoro-3-ethoxycarbonylphenylboronic acid
24901771
(3-ethoxycarbonyl-4-fluorophenyl)boronic acid
CCOC(=O)C1=C(F)C=CC(=C1)B(O)O
Melting Point 146.5°C to 148.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
Formula Weight 211.99
Percent Purity 95%
Chemical Name or Material 3-Ethoxycarbonyl-4-fluorophenylboronic acid
compliance-icons
Product Identifier
  • 3-Ethoxycarbonyl-4-fluorophenylboronic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.