missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-chlorophenyl)methanol, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
50 g
Specifications
Density | 1.2100g/mL |
Assay Percent Range | 97% |
Linear Formula | ClC6H4CH2OH |
Quantity | 10 g |
Formula Weight | 142.58 |
Percent Purity | 97% |
Chemical Name or Material | (3-Chlorophenyl)methanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'