Learn More
3-chloro-4-methylphenylboronic acid, 97%, Thermo Scientific Chemicals
Catalog Number p-4228523
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
175883-63-3
C7H8BClO2
170.40
MFCD04039010
YTJUYWRCAZWVSX-UHFFFAOYSA-N
3-chloro-4-methylphenyl boronic acid, 3-chloro-4-methylbenzeneboronic acid, 3-chloro-4-methylphenylboronicacid, 3-chloro-p-tolylboronic acid, boronic acid, 3-chloro-4-methylphenyl, pubchem5132, 3-chloro-4-methyl-phenyl boronic acid, acmc-209eaq, ksc489k9f, 3-chloro-4-methylphenyl-boronic acid
3854610
(3-chloro-4-methylphenyl)boronic acid
CC1=CC=C(C=C1Cl)B(O)O
175883-63-3
C7H8BClO2
170.40
MFCD04039010
YTJUYWRCAZWVSX-UHFFFAOYSA-N
3-chloro-4-methylphenyl boronic acid, 3-chloro-4-methylbenzeneboronic acid, 3-chloro-4-methylphenylboronicacid, 3-chloro-p-tolylboronic acid, boronic acid, 3-chloro-4-methylphenyl, pubchem5132, 3-chloro-4-methyl-phenyl boronic acid, acmc-209eaq, ksc489k9f, 3-chloro-4-methylphenyl-boronic acid
3854610
(3-chloro-4-methylphenyl)boronic acid
CC1=CC=C(C=C1Cl)B(O)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 228.3°C to 231.5°C
CAS Min % 96.0
Color White
CAS Max % 100.0
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder of Flakes
Assay Percent Range 96% min. (HPLC)
Formula Weight 170.4
Percent Purity 97%
Chemical Name or Material 3-chloro-4-methylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chloro-4-methylphenylboronic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.