missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluorobenzylamine, 97%, Thermo Scientific™

Catalog Number p-7047435
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
72235-55-3
C7H7ClFN
159.588
MFCD01631391
LYKWZKBLNOCJMA-UHFFFAOYSA-N
3-chloro-2-fluorobenzylamine, 3-chloro-2-fluorophenyl methanamine, 3-chloro-2-fluoro-benzylamine, 3-chloro-2-fluorophenyl methylamine, benzenemethanamine, 3-chloro-2-fluoro, 1-3-chloro-2-fluorophenyl methanamine, 2-fluoro-3-chlorobenzylamine, 2-fluoro-3-chlorobenzyl amine, 3-chloro-2-fluoro benzylamine, rarechem al bw 1387
2773614
(3-chloro-2-fluorophenyl)methanamine
C1=CC(=C(C(=C1)Cl)F)CN
This item is not returnable. View return policy
72235-55-3
C7H7ClFN
159.588
MFCD01631391
LYKWZKBLNOCJMA-UHFFFAOYSA-N
3-chloro-2-fluorobenzylamine, 3-chloro-2-fluorophenyl methanamine, 3-chloro-2-fluoro-benzylamine, 3-chloro-2-fluorophenyl methylamine, benzenemethanamine, 3-chloro-2-fluoro, 1-3-chloro-2-fluorophenyl methanamine, 2-fluoro-3-chlorobenzylamine, 2-fluoro-3-chlorobenzyl amine, 3-chloro-2-fluoro benzylamine, rarechem al bw 1387
2773614
(3-chloro-2-fluorophenyl)methanamine
C1=CC(=C(C(=C1)Cl)F)CN

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.5405
Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 159.59
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluorobenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'