Learn More
STEMCELL Technologies 3 cc Syringes
For accurate dispensing of methylcellulose-based medium
59.40 EUR - 122.85 EUR
Specifications
For Use With (Application) Colony Assay
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15680051
View Documents Promotion Details
STEMCELL Technologies
28240
100 Syringes/Bag
122.85 EUR
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 19-04-2024
to see stock.
17148071
View Documents Promotion Details
STEMCELL Technologies
28230
30 Syringes/Bag
59.40 EUR
Pack of 30
View alternative products
Save Up to 
»
Estimated Shipment: 19-04-2024
Description
  • Ideal for accurate dispensing of methylcellulose-based medium (e.g. MethoCult™), when used with Blunt-End Needles (Catalog #28110) purchased from STEMCELL Technologies to ensure compatibility and accuracy
Specifications
Specifications
Colony Assay