missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(4-ethylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64008.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458636

  • 253.00 EUR / 1g


Estimated Shipment date
from Supplier 24-04-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27665242

Specifications

3-Bromo-N-(4-ethylphenyl)-5-(trifluoromethyl)benzenesulfonamide
MFCD27665242
Solid
1g
408.23
97%
Videos
SDS