missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(3-chloro-4-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64719.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15459106

  • 949.00 EUR / 5g


Estimated Shipment date
from Supplier 24-04-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27665307

Specifications

3-Bromo-N-(3-chloro-4-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
MFCD27665307
Solid
5g
428.65
97%
Videos
SDS