Learn More
Alfa Aesar™ 3-Bromo-N-(2-methoxybenzyl)-5-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H64107.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15458706
  • 891.00 EUR / 5g
Estimated Shipment date
from Supplier 18-06-2019
Add to basket
Specifications
Chemical Identifiers
MFCD27975665
Specifications
3-Bromo-N-(2-methoxybenzyl)-5-(trifluoromethyl)benzenesulfonamide
MFCD27975665
Solid
5g
424.23
97%