missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2,5-dimethoxyphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64638.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459046

  • 176.00 EUR / 1g


Estimated Shipment date
from Supplier 13-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27665299

Specifications

3-Bromo-N-(2,5-dimethoxyphenyl)-5-(trifluoromethyl)benzenesulfonamide
MFCD27665299
Solid
1g
440.23
97%
Videos
SDS