missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2,5-difluorophenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64550.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15458976

  • 899.00 EUR / 5g


Estimated Shipment date
from Supplier 19-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27665287

Specifications

3-Bromo-N-(2,5-difluorophenyl)-5-(trifluoromethyl)benzenesulfonamide
MFCD27665287
Solid
5g
416.16
97%
Videos
SDS