missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-5-(3,5-dimethylphenylcarbamoyl)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H61931.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15446866

  • 900.00 EUR / 1g


Estimated Shipment date
from Supplier 20-06-2019
Add to basket

Specifications

Chemical Identifiers

MFCD26143302

Specifications

3-Bromo-5-(3,5-dimethylphenylcarbamoyl)benzeneboronic acid
MFCD26143302
Solid
1g
349.01
97%
Videos
SDS