missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-6-chloropyridazine, 97%, Thermo Scientific Chemicals

Catalog Number p-4812908
Click to view available options
Quantity:
50 g
100 g
5469-69-2
C4H4ClN3
129.55
MFCD00051506
DTXVKPOKPFWSFF-UHFFFAOYSA-N
3-amino-6-chloropyridazine, 3-pyridazinamine, 6-chloro, 6-chloro-3-pyridazinamine, 6-amino-3-chloropyridazine, 6-chloro-3-aminopyridazine, 6-chloropyridazin-3-ylamine, 6-chloro-pyridazin-3-ylamine, unii-ijs443slzg, 3-chloro-6-pyridazinamine, 6-chloro-3-pyridazinylamine
21643
6-chloropyridazin-3-amine
NC1=CC=C(Cl)N=N1
5469-69-2
C4H4ClN3
129.55
MFCD00051506
DTXVKPOKPFWSFF-UHFFFAOYSA-N
3-amino-6-chloropyridazine, 3-pyridazinamine, 6-chloro, 6-chloro-3-pyridazinamine, 6-amino-3-chloropyridazine, 6-chloro-3-aminopyridazine, 6-chloropyridazin-3-ylamine, 6-chloro-pyridazin-3-ylamine, unii-ijs443slzg, 3-chloro-6-pyridazinamine, 6-chloro-3-pyridazinylamine
21643
6-chloropyridazin-3-amine
NC1=CC=C(Cl)N=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 210.0°C to 211.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 50 g
Formula Weight 129.55
Percent Purity 97%
Chemical Name or Material 3-Amino-6-chloropyridazine
compliance-icons
Product Identifier
  • 3-Amino-6-chloropyridazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'