Learn More
3-Amino-5-ethyl-1H-pyrazole, 97%, Thermo Scientific Chemicals
Catalog Number p-7077310
Click to view available options
Quantity:
1 g
5 g
1904-24-1
C5H9N3
111.15
MFCD08060982
AXDGPQLEVYSXNL-UHFFFAOYSA-N
3-ethyl-1h-pyrazol-5-amine, 5-amino-3-ethyl-1h-pyrazole, 3-amino-5-ethylpyrazole, 3-amino-5-ethyl-1h-pyrazole, 5-ethyl-2h-pyrazol-3-ylamine, 1h-pyrazol-3-amine, 5-ethyl, 5-ethyl-2h-pyrazol-3-amine, ethyl-5-aminopyrazole, 3-amino-5-ethypyrazole, 1h-pyrazol-5-amine, 3-ethyl
21991095
5-ethyl-1H-pyrazol-3-amine
CCC1=CC(N)=NN1
1904-24-1
C5H9N3
111.15
MFCD08060982
AXDGPQLEVYSXNL-UHFFFAOYSA-N
3-ethyl-1h-pyrazol-5-amine, 5-amino-3-ethyl-1h-pyrazole, 3-amino-5-ethylpyrazole, 3-amino-5-ethyl-1h-pyrazole, 5-ethyl-2h-pyrazol-3-ylamine, 1h-pyrazol-3-amine, 5-ethyl, 5-ethyl-2h-pyrazol-3-amine, ethyl-5-aminopyrazole, 3-amino-5-ethypyrazole, 1h-pyrazol-5-amine, 3-ethyl
21991095
5-ethyl-1H-pyrazol-3-amine
CCC1=CC(N)=NN1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 111.15
Percent Purity 97%
Chemical Name or Material 3-Amino-5-ethyl-1H-pyrazole
compliance-icons
Product Identifier
  • 3-Amino-5-ethyl-1H-pyrazole, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.