Learn More
3-Amino-2-cyclohexen-1-one, 95%, Thermo Scientific™
Catalog Number p-4812899
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
5220-49-5
C6H9NO
111.14
MFCD00013783
ZZMRPOAHZITKBV-UHFFFAOYSA-N
3-amino-2-cyclohexen-1-one, 3-aminocyclohex-2-enone, 2-cyclohexen-1-one, 3-amino, unii-v596547zke, 3-amino-2-cyclohexen-1-ketone, 3-amino-2-cyclohexenone, 3-aminocyclohexenone, 3-aminocyclohex-enone, pubchem14449, acmc-209kyd
78892
3-aminocyclohex-2-en-1-one
NC1=CC(=O)CCC1
5220-49-5
C6H9NO
111.14
MFCD00013783
ZZMRPOAHZITKBV-UHFFFAOYSA-N
3-amino-2-cyclohexen-1-one, 3-aminocyclohex-2-enone, 2-cyclohexen-1-one, 3-amino, unii-v596547zke, 3-amino-2-cyclohexen-1-ketone, 3-amino-2-cyclohexenone, 3-aminocyclohexenone, 3-aminocyclohex-enone, pubchem14449, acmc-209kyd
78892
3-aminocyclohex-2-en-1-one
NC1=CC(=O)CCC1
Melting Point 133.0°C to 135.0°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 111.14
Percent Purity 95%
Chemical Name or Material 3-Amino-2-cyclohexen-1-one
compliance-icons
Product Identifier
  • 3-Amino-2-cyclohexen-1-one
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.