missing translation for 'onlineSavingsMsg'
Learn More

3-Amidinopyridine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7025081
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
7356-60-7
C6H8ClN3
157.601
MFCD00054326
MKJPBOVLAZADQJ-UHFFFAOYSA-N
nicotinimidamide hydrochloride, 3-amidinopyridine hydrochloride, pyridine-3-carboximidamide hydrochloride, nicotinamidine hydrochloride, 3-pyridinecarboximidamide, monohydrochloride, nicotinamidine, hydrochloride, pyridine-3-carbonamidine hydrochloride, pyridine-3-carboxamidine hydrochloride, 3-pyridinecarboxamidine hydrochloride, pyridine-3-carboximidamide hcl
202042
pyridine-3-carboximidamide;hydrochloride
C1=CC(=CN=C1)C(=N)N.Cl
This item is not returnable. View return policy
7356-60-7
C6H8ClN3
157.601
MFCD00054326
MKJPBOVLAZADQJ-UHFFFAOYSA-N
nicotinimidamide hydrochloride, 3-amidinopyridine hydrochloride, pyridine-3-carboximidamide hydrochloride, nicotinamidine hydrochloride, 3-pyridinecarboximidamide, monohydrochloride, nicotinamidine, hydrochloride, pyridine-3-carbonamidine hydrochloride, pyridine-3-carboxamidine hydrochloride, 3-pyridinecarboxamidine hydrochloride, pyridine-3-carboximidamide hcl
202042
pyridine-3-carboximidamide;hydrochloride
C1=CC(=CN=C1)C(=N)N.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 187°C to 189°C
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 157.6
Percent Purity 97%
Chemical Name or Material 3-Amidinopyridine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'