missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethyl-4-isoxazolecarbaldehyde, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
10 g
Specifica
Assay Percent Range | 97% |
Quantity | 250 mg |
Formula Weight | 125.13 |
Percent Purity | 97% |
Chemical Name or Material | 3, 5-Dimethyl-4-isoxazolecarbaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'