missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluoro-N-(4-fluoro-2-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H61245.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15446386

  • 646.00 EUR / 1g


Estimated Shipment date
from Supplier 24-04-2019
Add to basket

Specifications

Chemical Identifiers

MFCD07264129

Specifications

3,5-Difluoro-N-(4-fluoro-2-methylphenyl)benzamide
MFCD07264129
Solid
1g
265.23
97%
Videos
SDS