missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluoro-N-(2-fluoro-5-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H61212.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15446356

  • 720.00 EUR / 1g


Estimated Shipment date
from Supplier 19-06-2019
Add to basket

Specifications

Chemical Identifiers

MFCD08481408

Specifications

3,5-Difluoro-N-(2-fluoro-5-methylphenyl)benzamide
MFCD08481408
Solid
1g
265.23
97%
Videos
SDS