Learn More
Alfa Aesar™ 3-(4-Oxo-2-thioxo-3-thiazolidinyl)benzoic acid, 95%
Brand:  Alfa Aesar™ H63979.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15458616
  • 1291.00 EUR / 5g
Estimated Shipment date
from Supplier 03-09-2018
Add to basket
Specifications
Chemical Identifiers
3-(4-Oxo-2-thioxo-3-thiazolidinyl)benzoic acid
MFCD01184188
Specifications
3-(4-Oxo-2-thioxo-3-thiazolidinyl)benzoic acid
MFCD01184188
Solid
95%
253.29
5g