missing translation for 'onlineSavingsMsg'
Learn More

3,4-Methylenedioxy-beta-nitrostyrene, 98%, Thermo Scientific™

Catalog Number p-7045022
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
1485-00-3
C9H7NO4
193.158
MFCD00014575
KFLWBZPSJQPRDD-ONEGZZNKSA-N
3,4-methylenedioxy-beta-nitrostyrene, mns, syk inhibitor iii, 1,3-benzodioxole, 5-nitrovinyl, 5-2-nitrovinyl benzo d 1,3 dioxole, 1,3-benzodioxole, 5-2-nitroethenyl, 5-2-nitrovinyl-1,3-benzodioxole, 5-2-nitroethenyl-1,3-benzodioxole, styrene, 3,4-methylenedioxy-beta-nitro, e-5-2-nitrovinyl benzo d 1,3 dioxole
672296
5-[(E)-2-nitroethenyl]-1,3-benzodioxole
C1OC2=C(O1)C=C(C=C2)C=C[N+](=O)[O-]
This item is not returnable. View return policy
1485-00-3
C9H7NO4
193.158
MFCD00014575
KFLWBZPSJQPRDD-ONEGZZNKSA-N
3,4-methylenedioxy-beta-nitrostyrene, mns, syk inhibitor iii, 1,3-benzodioxole, 5-nitrovinyl, 5-2-nitrovinyl benzo d 1,3 dioxole, 1,3-benzodioxole, 5-2-nitroethenyl, 5-2-nitrovinyl-1,3-benzodioxole, 5-2-nitroethenyl-1,3-benzodioxole, styrene, 3,4-methylenedioxy-beta-nitro, e-5-2-nitrovinyl benzo d 1,3 dioxole
672296
5-[(E)-2-nitroethenyl]-1,3-benzodioxole
C1OC2=C(O1)C=C(C=C2)C=C[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 159°C to 163°C
Quantity 10 g
Beilstein 192350
Sensitivity Light sensitive
Formula Weight 193.16
Percent Purity 98%
Chemical Name or Material 3,4-Methylenedioxy-beta-nitrostyrene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'