missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzylamine, 98%, Thermo Scientific Chemicals

Catalog Number p-104522
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
102-49-8
C7H7Cl2N
176.05
MFCD00008114
IXHNFOOSLAWRBQ-UHFFFAOYSA-N
3,4-dichlorobenzylamine, 3,4-dichlorophenyl methanamine, benzenemethanamine, 3,4-dichloro, 3,4-dichloro-benzylamine, benzylamine, 3,4-dichloro, 3,4-dichlorophenyl methylamine, 1-3,4-dichlorophenyl methanamine, 3,4-dichlorophenyl methyl amine, 3,4-dichlorobezylamine
1608
(3,4-dichlorophenyl)methanamine
C1=CC(=C(C=C1CN)Cl)Cl
This item is not returnable. View return policy
102-49-8
C7H7Cl2N
176.05
MFCD00008114
IXHNFOOSLAWRBQ-UHFFFAOYSA-N
3,4-dichlorobenzylamine, 3,4-dichlorophenyl methanamine, benzenemethanamine, 3,4-dichloro, 3,4-dichloro-benzylamine, benzylamine, 3,4-dichloro, 3,4-dichlorophenyl methylamine, 1-3,4-dichlorophenyl methanamine, 3,4-dichlorophenyl methyl amine, 3,4-dichlorobezylamine
1608
(3,4-dichlorophenyl)methanamine
C1=CC(=C(C=C1CN)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3200g/mL
Boiling Point 80°C to 82°C (0.6 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3CH2NH2
Refractive Index 1.5775 to 1.5795
Quantity 5 mL
Specific Gravity 1.32
Formula Weight 176.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3, 4-Dichlorobenzylamine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dichlorobenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'