Learn More
3,4,5-Trimethoxybenzyl alcohol, 98%, Thermo Scientific Chemicals
Catalog Number 11454384
Click to view available options
Quantity:
10 g
50 g
250 g
3840-31-1
C10H14O4
198.218
MFCD00004639
QPHLRCUCFDXGLY-UHFFFAOYSA-N
3,4,5-trimethoxybenzyl alcohol, 3,4,5-trimethoxyphenyl methanol, benzenemethanol, 3,4,5-trimethoxy, 3,4,5-trimethoxybenzylalcohol, 3,4,5-trimethoxybenzylic alcohol, benzyl alcohol, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methan-1-ol, acmc-1cp7y, 3,4,5-trimethoxybenzalcohol, ksc494s0n
77449
(3,4,5-trimethoxyphenyl)methanol
COC1=CC(=CC(=C1OC)OC)CO
3840-31-1
C10H14O4
198.218
MFCD00004639
QPHLRCUCFDXGLY-UHFFFAOYSA-N
3,4,5-trimethoxybenzyl alcohol, 3,4,5-trimethoxyphenyl methanol, benzenemethanol, 3,4,5-trimethoxy, 3,4,5-trimethoxybenzylalcohol, 3,4,5-trimethoxybenzylic alcohol, benzyl alcohol, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methan-1-ol, acmc-1cp7y, 3,4,5-trimethoxybenzalcohol, ksc494s0n
77449
(3,4,5-trimethoxyphenyl)methanol
COC1=CC(=CC(=C1OC)OC)CO
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 1.199
Boiling Point 228°C (25 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5445
Quantity 10 g
Beilstein 2104523
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material 3,4,5-Trimethoxybenzyl alcohol