missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diallylbisphenol A diacetate, epoxy curative, EC-392, Thermo Scientific Chemicals

Catalog Number p-7080097
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
1071466-61-9
C25H28O4
392.495
MFCD22200539
GJTZQXNRNNDGRG-UHFFFAOYSA-N
2,2'-diallyl bisphenol a diacetate ether, 4,4'-isopropylidene bis 2-allylphenol acetate, phenyl ester epoxy curative hybrid of bisphenol a, 3,3'-diallylbisphenol a diacetate, epoxy curative, ec-392, 4-2-4-acetyloxy-3-prop-2-en-1-yl phenyl propan-2-yl-2-prop-2-en-1-yl phenyl acetate
59215472
[4-[2-(4-acetyloxy-3-prop-2-enylphenyl)propan-2-yl]-2-prop-2-enylphenyl] acetate
CC(=O)OC1=C(C=C(C=C1)C(C)(C)C2=CC(=C(C=C2)OC(=O)C)CC=C)CC=C
This item is not returnable. View return policy
1071466-61-9
C25H28O4
392.495
MFCD22200539
GJTZQXNRNNDGRG-UHFFFAOYSA-N
2,2'-diallyl bisphenol a diacetate ether, 4,4'-isopropylidene bis 2-allylphenol acetate, phenyl ester epoxy curative hybrid of bisphenol a, 3,3'-diallylbisphenol a diacetate, epoxy curative, ec-392, 4-2-4-acetyloxy-3-prop-2-en-1-yl phenyl propan-2-yl-2-prop-2-en-1-yl phenyl acetate
59215472
[4-[2-(4-acetyloxy-3-prop-2-enylphenyl)propan-2-yl]-2-prop-2-enylphenyl] acetate
CC(=O)OC1=C(C=C(C=C1)C(C)(C)C2=CC(=C(C=C2)OC(=O)C)CC=C)CC=C

It is used in epoxy systems or in combination with acrylates, methacrylates, and maleimides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Recommended for use in epoxy systems or in combination with acrylates, methacrylates, and maleimides
TRUSTED_SUSTAINABILITY
Color Amber
Physical Form Amber liquid
Quantity 25 g
Formula Weight 392.5
Chemical Name or Material 3,3'-Diallylbisphenol A diacetate, Epoxy curative, EC-392

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'